missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LPIN1 Monoclonal Antibody (OTI4C11), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA806417
This item is not returnable.
View return policy
Description
This gene represents a candidate gene for human lipodystrophy, characterized by loss of body fat, fatty liver, hypertriglyceridemia, and insulin resistance. Mouse studies suggest that this gene functions during normal adipose tissue development and may also play a role in human triglyceride metabolism.Specifications
| LPIN1 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q14693 | |
| LPIN1 | |
| Human recombit protein fragment corresponding to amino acids 1-274 of human LPIN1 produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b |
| Immunohistochemistry (Paraffin) | |
| OTI4C11 | |
| Unconjugated | |
| LPIN1 | |
| PAP1 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 23175 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction