Learn More
LPIN1 Monoclonal Antibody (OTI1B2), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand:  Origene Technologies TA806208
Encompass_Preferred
 View more versions of this product
Product Code. 17586293
  • € 540.87 / 100µL
Description
This gene represents a candidate gene for human lipodystrophy, characterized by loss of body fat, fatty liver, hypertriglyceridemia, and insulin resistance. Mouse studies suggest that this gene functions during normal adipose tissue development and may also play a role in human triglyceride metabolism.
Specifications
Specifications
LPIN1
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Q14693
LPIN1
Human recombit protein fragment corresponding to amino acids 1-274 of human LPIN1 produced in E.coli.
100 μL
Primary
Human
Antibody
IgG1
Immunohistochemistry (Paraffin), Western Blot
OTI1B2
Unconjugated
LPIN1
PAP1
Mouse
Affinity Chromatography
RUO
23175
-20° C, Avoid Freeze/Thaw Cycles