missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ LPIN2 Polyclonal Antibody
GREENER_CHOICE

Product Code. 16587931
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16587931 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16587931 Supplier Invitrogen™ Supplier No. PA590683

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: TCDVGVSSDD DKGAQAARGS SNASLKEEEC KEPLLFHSGD HYPLSDGDWS PLETTYPQTA CPKSDSELEV KPAESLLRSE SHMEWTWGGF PESTKVSKRE RSDHHPRTAT ITPSENTHFR VIPSEDNLIS EVEKDASMED TVCTIVKPKP RALGTQMSDP TSVAELLEPP LESTQISSML DADHLPNAAL AEAPSESKPA AKVDSPSKKK GVHKRSQHQG P; Positive Samples: LO2, 293T, A-549, DU145, Mouse liver, Mouse lung, Mouse kidney, Mouse spleen; Cellular Location: Cytoplasm, Endoplasmic reticulum membrane, Nucleus, cytosol.

Plays important roles in controlling the metabolism of fatty acids at differents levels. Acts as a magnesium-dependent phosphatidate phosphatase enzyme which catalyzes the conversion of phosphatidic acid to diacylglycerol during triglyceride, phosphatidylcholine and phosphatidylethanolamine biosynthesis in the reticulum endoplasmic membrane. Acts also as a nuclear transcriptional coactivator for PPARGC1A to modulate lipid metabolism.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LPIN2
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.57 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene LPIN2
Gene Accession No. Q92539, Q99PI5
Gene Alias 2610511G02Rik; AI481352; AW742896; KIAA0249; lipin 2; lipin-2; Lpin2; Phosphatidate phosphatase LPIN2
Gene Symbols LPIN2
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 180-400 of human LPIN2 (NP_0554611).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 64898, 9663
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.