missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LPIN3 Monoclonal Antibody (OTI1C9), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA805038S
This item is not returnable.
View return policy
Description
LPIN3 Monoclonal antibody specifically detects LPIN3 in Human samples. It is validated for Immunohistochemistry (Paraffin)Specifications
| LPIN3 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q9BQK8 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 64900 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG2b |
| Immunohistochemistry (Paraffin) | |
| OTI1C9 | |
| Unconjugated | |
| LPIN3 | |
| 9130206L11Rik; AA438110; AV236139; dJ450M14.2; dJ450M14.3; dJ620E11.2; lipin 3; lipin 3-like; lipin-3; Lipin-3-like; LIPN3L; Lpin3; mKIAA4023; Phosphatidate phosphatase LPIN3; RP4-620E11.3; SMP2 | |
| Full length human recombinant protein of human LPIN3 produced in HEK293T cell. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction