Learn More
Invitrogen™ LPL Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA585126
Description
Keep as concentrated solution. Predicted reactivity: Mouse (90%), Rat (90%), Cat (94%), Pig (93%), Rabbit (92%), Sheep (91%), Bovine (91%), Guinea pig (86%). Positive Control: Molt-4, NIH-3T3. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
LPL encodes lipoprotein lipase, which is expressed in heart, muscle, and adipose tissue. LPL functions as a homodimer, and has the dual functions of triglyceride hydrolase and ligand/bridging factor for receptor-mediated lipoprotein uptake. Severe mutations that cause LPL deficiency result in type I hyperlipoproteinemia, while less extreme mutations in LPL are linked to many disorders of lipoprotein metabolism.
Specifications
| LPL | |
| Polyclonal | |
| Unconjugated | |
| LPL | |
| adipose lipoprotein lipase; alpha lipase; HDLCQ11; I79_006077; LIPD; lipoprotein lipase; lipoprotein lipase (EC 3.1.1.34); lipoprotein lipase precursor; lipoprotein lipase preprotein; Lipoprotein lipase-like protein; LPL; O 1-4-5; phospholipase A1; triacylglycerol lipase | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 16956, 396219, 4023, 443408 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.11 mg/mL | |
| PBS with 1% BSA, 20% glycerol and 0.025% ProClin 300; pH 7 | |
| P06858, P11152, P11602, Q29524 | |
| LPL | |
| Recombinant protein encompassing a sequence within the center region of human LPL. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Sheep, Chicken | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.