Learn More
Invitrogen™ Lplunc1 Monoclonal Antibody (1F11E3)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA517125
Description
MA5-17125 targets Lplunc1 in indirect ELISA and WB applications and shows reactivity with Mouse samples. The MA5-17125 immunogen is purified recombinant fragment of mouse Lplunc1 (amino acids: 248-475) expressed in E. Coli. MA5-17125 detects Lplunc1 which has a predicted molecular weight of approximately 52.4kDa.
The protein encoded by this gene may be involved in the innate immune response to bacterial exposure in the mouth, nasal cavities, and lungs. The encoded protein is secreted and is a member of the BPI/LBP/PLUNC protein superfamily. This gene is found with other members of the superfamily in a cluster on chromosome 20.
Specifications
| Lplunc1 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 0.05% sodium azide | |
| Q61114 | |
| BPIFB1 | |
| Purified recombinant fragment of mouse Lplunc1 (amino acids: 248-475) expressed in E. Coli. | |
| 100 μg | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG1 |
| ELISA, Western Blot | |
| 1F11E3 | |
| Unconjugated | |
| BPIFB1 | |
| BPI fold containing family B member 1; BPI fold containing family B, member 1; BPI fold-containing family B member 1; BPIFB1; C20orf114; long palate, lung and nasal epithelium carcinoma associated 1; long palate, lung and nasal epithelium carcinoma-associated protein 1; Lplunc1; UNQ706/PRO1357; VEMSGP; von Ebner minor salivary gland protein; von Ebner minor salivary protein | |
| Mouse | |
| Protein G | |
| RUO | |
| 228801 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.