Learn More
Invitrogen™ LPP Monoclonal Antibody (8B3A11)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA515379
Description
MA5-15379 targets LPP in indirect ELISA, IF, IHC, and WB applications and shows reactivity with Hamster, Human, mouse, and Non-human primate samples. The MA5-15379 immunogen is purified recombinant fragment of human LPP expressed in E. Coli. MA5-15379 detects LPP which has a predicted molecular weight of approximately 66kDa.
This gene encodes a member of a subfamily of LIM domain proteins that are characterized by an N-terminal proline rich region and three C-terminal LIM domains. The encoded protein localizes to the cell periphery in focal adhesions and may be involved in cell-cell adhesion and cell motility. This protein also shuttles through the nucleus and may function as a transcriptional co-activator. This gene is located at the junction of certain disease related chromosomal translocations which result in the expression of chimeric proteins that may promote tumor growth. Alternate splicing results in multiple transcript variants.
Specifications
| LPP | |
| Monoclonal | |
| Conc. Not Determined | |
| Ascites with 0.03% sodium azide | |
| Q8BFW7, Q93052 | |
| LPP | |
| Purified recombinant fragment of human LPP expressed in E. Coli. | |
| RUO | |
| 100753874, 210126, 4026 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 8B3A11 | |
| Unconjugated | |
| LPP | |
| 9430020K16Rik; AA959454; AU024130; B130055L10Rik; C79715; D630048H16; I79_000864; LIM domain containing preferred translocation partner in lipoma; LIM domain-containing preferred translocation partner in lipoma; LIM protein; lipoma preferred partner; Lipoma-preferred partner; Lipoma-preferred partner homolog; Lipoma-preferred-partner; LPP | |
| Mouse | |
| 100 μL | |
| Primary | |
| Human, Mouse, Monkey, Hamster | |
| Antibody | |
| IgG1 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.