missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LpxT Antibody, Alexa Fluor™ 405, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-23512AF405
This item is not returnable.
View return policy
Description
LpxT Polyclonal antibody specifically detects LpxT in E. coli samples. It is validated for Western Blot, ELISA
Specifications
| LpxT | |
| Polyclonal | |
| Western Blot, ELISA | |
| ECK2168, Kdo(2)-lipid A phosphotransferase, Lipid A 1-diphosphate synthase, lipid A-core phosphotransferase, Undecaprenyl pyrophosphate:lipid A 1-phosphate phosphotransferase, yeiU | |
| Antibody was raised against an 18 amino acid peptide near the carboxy terminus of E. coli LpxT. The immunogen is located within the last 50 amino acids of LpxT. | |
| 0.1 mL | |
| Primary | |
| E. coli | |
| Purified |
| Western Blot, ELISA | |
| Alexa Fluor 405 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 946693 | |
| Store at 4C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction