missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LRAT Antibody (M34-P1F10) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
€ 201.60 - € 372.75
Specifications
| Antigen | LRAT |
|---|---|
| Clone | M34-P1F10 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:100 - 1:2000, ELISA 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/ Immunofluorescence 1:10 - 1:500 |
| Applications | Western Blot, ELISA, Immunohistochemistry, Immunofluorescence |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18614578
|
Novus Biologicals
NBP2-50444-0.025ml |
0.025 mL |
€ 214.00 € 201.60 / 0.03mL Save € 12.40 5% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18603029
|
Novus Biologicals
NBP2-50444 |
0.1 mL |
€ 395.00 € 372.75 / 0.10mL Save € 22.25 5% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
LRAT Monoclonal antibody specifically detects LRAT in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ ImmunofluorescenceSpecifications
| LRAT | |
| 1 mg/mL | |
| Western Blot, ELISA, Immunohistochemistry, Immunofluorescence | |
| Unconjugated | |
| Mouse | |
| Endocrinology, Neuroscience, Signal Transduction | |
| PBS | |
| 9227 | |
| IgG1 | |
| Protein A purified |
| M34-P1F10 | |
| Western Blot 1:100 - 1:2000, ELISA 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/ Immunofluorescence 1:10 - 1:500 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| EC 2.3.1.135, LCA14, lecithin retinol acyltransferase, lecithin retinol acyltransferase (phosphatidylcholine--retinolO-acyltransferase), MGC33103, Phosphatidylcholine--retinol O-acyltransferase | |
| Peptide Sequence - RDQRSVLASA (amino acids 190 -199) | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title