missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ LRCH1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA554441
This item is not returnable.
View return policy
Description
Immunogen sequence: LHQHVEDGKK DSDSGVGSDN GDKRLSATEP SDEDTVSLNV PMSNIMEEEQ IIKEDSCHRL SPVKGEFHQE FQPEPSLLGD STNSGEERDQ FTDRADGLHS EFMNYKATTA EDC Highest antigen sequence identity to the following orthologs: Mouse - 74%, Rat - 74%.
This gene encodes a protein with a leucine-rich repeat and a calponin homology domain. Polymorphism in this gene may be associated with susceptibility to knee osteoarthritis. Alternative splicing results in multiple transcript variants encoding different isoforms.
Specifications
| LRCH1 | |
| Polyclonal | |
| Unconjugated | |
| LRCH1 | |
| Calponin homology domain-containing protein 1; CHDC1; KIAA1016; leucine rich repeats and calponin homology domain containing 1; leucine-rich repeat and calponin homology domain-containing protein 1; leucine-rich repeats and calponin homology (CH) domain containing 1; LRCH1; Neuronal protein 81; NP81 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 23143 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9Y2L9 | |
| LRCH1 | |
| Recombinant protein corresponding to Human LRCH1. Recombinant protein control fragment (Product #RP-90244). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction