missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ LRCH2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5145148
This item is not returnable.
View return policy
Description
Immunogen sequence: RNKPKQTVEC EKSVSADEVN SPLSPLTWQP LENQKDQIDE QPWPESHPII WQSEERRRSK QIRKEYFKYK SMRKSSSGNE NDEQDSDNAN MSTQSPVS Antigen sequence identity: Mouse 85%; Rat 86%.
Leucine rich repeat and calponin homology domain containing protein 2 (LRCH2) also named as KIAA1495 is a 765 amino acid protein, which contains one CH domain and nine LRR repeats. The function of LRCH2 is still non-known.
Specifications
| LRCH2 | |
| Polyclonal | |
| Unconjugated | |
| LRCH2 | |
| AI316885; dA204F4.4; Kiaa1495; leucine rich repeats and calponin homology domain containing 2; leucine-rich repeat and calponin homology domain-containing protein 2; leucine-rich repeats and calponin homology (CH) domain containing 2; Lrch2; mKIAA1495 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 57631 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid | |
| IgG |
| Immunocytochemistry | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q5VUJ6 | |
| LRCH2 | |
| Recombinant Protein Epitope Signature Tag (PrEST) antigen sequence. Antigen range: 498-595. Recombinant protein control fragment (Product #RP-109954). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction