missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ LRCH3 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals™ NBP3-29698
This item is not returnable.
View return policy
Description
LRCH3 Polyclonal antibody specifically detects LRCH3 in Human samples. It is validated for Western Blot
Specifications
| LRCH3 | |
| Polyclonal | |
| Tris-citrate/phosphate pH 7 -8. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 84859 | |
| Store at 4°C. Do not freeze. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| FLJ20994, FLJ43245, FLJ44976,2210409B11Rik, leucine-rich repeat and calponin homology domain-containing protein 3, leucine-rich repeats and calponin homology (CH) domain containing 3, MGC4126 | |
| Between 500 and 550 | |
| 100 μg | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction