missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ LRCH3 Polyclonal Antibody

Product Code. 15760881 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15760881 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15760881 Supplier Invitrogen™ Supplier No. PA552933

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: HASPLPPSAA PTTDSTDSIT GQNSRQREEE LELIDQLRKH IEYRLKVSLP CDLGAALTDG VVLCHLANHV RPRSVPSIHV PSP Highest antigen sequence identity to the following orthologs: Mouse - 86%, Rat - 84%.

LRCH3 (Leucine Rich Repeats And Calponin Homology Domain Containing 3) is a Protein Coding gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LRCH3
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.30 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Lrch3
Gene Accession No. Q8BVU0, Q96II8
Gene Alias 2210409B11Rik; AW215594; DISP complex protein LRCH3; Gm1742; leucine rich repeats and calponin homology domain containing 3; leucine-rich repeat and calponin homology domain-containing protein 3; leucine-rich repeats and calponin homology (CH) domain containing 3; LRCH3; MGC4126
Gene Symbols Lrch3
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human LRCH3. Recombinant protein control fragment (Product #RP-90560).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 70144, 84859
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.