Learn More
LRFN5 Rabbit, anti-Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 26856-1-AP-20UL
 View more versions of this product
Product Code. 18049655
  • € 145.00 / 20µL
Description
The LRFN5 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human LRFN5. This antibody recognizes mouse, rat antigen. The LRFN5 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
Specifications
LRFN5
350 μg/mL
Rabbit
Primary
Mouse, Rat
150 kDa
IgG
Western Blot, ELISA
C14orf146, FIGLER8, LRFN5, SALM5
20 μL
145581
Store at -20°C. Stable for one year after shipment.