missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ LRG1 Monoclonal Antibody (C2), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen™ MA550924
This item is not returnable.
View return policy
Description
The leucine-rich repeat (LRR) family of proteins, including LRG1, have been shown to be involved in protein-protein interaction, signal transduction, and cell adhesion and development. LRG1 is expressed during granulocyte differentiation.
Specifications
| LRG1 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.05% Proclin 300; pH 7.4 | |
| P02750 | |
| LRG1 | |
| Recombinant Human LRG1 protein, Thr37-Leu340 (Accession #P02750). | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 κ |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| C2 | |
| Unconjugated | |
| LRG1 | |
| 1300008B03Rik; 2310031E04Rik; A2GL; HMFT1766; leucine rich alpha 2 glycoprotein; leucine rich alpha-2-glycoprotein 1; leucine-rich alpha-2-glycoprotein; leucine-rich alpha-2-glycoprotein 1; Leucine-rich alpha-2-glycoprotein precursor; LRG; LRG 1; LRG1; LRG-1; Lrhg | |
| Mouse | |
| Protein A/G | |
| RUO | |
| 116844 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction