Learn More
Invitrogen™ LRG1 Recombinant Rabbit Monoclonal Antibody (24GB545)
GREENER_CHOICE
Rabbit Recombinant Monoclonal Antibody
Brand:  Invitrogen™ MA557438
 View more versions of this product
Product Code. 30275169
  • € 580.00 / 100µL
Description
LRG1 Recombinant Monoclonal Antibody for Western Blot, ICC/IF
The leucine-rich repeat (LRR) family of proteins, including LRG1, have been shown to be involved in protein-protein interaction, signal transduction, and cell adhesion and development. LRG1 is expressed during granulocyte differentiation.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
LRG1
Recombinant Monoclonal
Unconjugated
LRG1
1300008B03Rik; 2310031E04Rik; A2GL; HMFT1766; leucine rich alpha 2 glycoprotein; leucine rich alpha-2-glycoprotein 1; leucine-rich alpha-2-glycoprotein; leucine-rich alpha-2-glycoprotein 1; Leucine-rich alpha-2-glycoprotein precursor; LRG; LRG 1; LRG1; LRG-1; Lrhg
Rabbit
Affinity chromatography
RUO
116844
-20°C
Liquid
Western Blot, Immunocytochemistry
24GB545
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
P02750
LRG1
A synthesized peptide derived from human LRG1 (aa 250-300).