missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ LRGUK Polyclonal Antibody
GREENER_CHOICE

Product Code. 15714091 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15714091 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15714091 Supplier Invitrogen™ Supplier No. PA556175

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: SNMGDFLHST DRNYLIKFWA KLSAKKTPAE RDSIHRQHEA ARQALMGRIR PDHTLLFQRG PVPAPLTSGL HYYTTLEELW KSFDLCEDYF KPPFG Highest antigen sequence identity to the following orthologs: Mouse - 64%, Rat - 61%.

The LRGUK (Leucine-Rich Repeat and Guanylate Kinase-Like Domain Containing 1) gene encodes a protein that plays a critical role in male fertility. It is particularly important for the proper function of the manchette, a microtubule-based structure involved in sperm head shaping and tail formation during spermiogenesis. Research has shown that LRGUK1 is part of a multiprotein complex essential for the initiation of sperm tail growth from the centriole/basal body. Mutations or disruptions in the LRGUK1 gene can lead to defective manchette function, resulting in abnormal sperm morphology and infertility. This gene's role in spermatogenesis underscores its importance in reproductive health, with ongoing studies aimed at elucidating its specific interactions and mechanisms within the manchette complex.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LRGUK
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene LRGUK
Gene Accession No. Q96M69
Gene Alias 4921528H16Rik; CFAP246; leucine rich repeats and guanylate kinase domain containing; leucine-rich repeat and guanylate kinase domain-containing protein; leucine-rich repeats and guanylate kinase domain containing; Lrguk; RGD1308226
Gene Symbols LRGUK
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human LRGUK. Recombinant protein control fragment (Product #RP-92765).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 136332
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.