missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LRH-1/NR5A2 Antibody (OTI3H8), FITC, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-72521F
This item is not returnable.
View return policy
Description
LRH-1/NR5A2 Monoclonal specifically detects LRH-1/NR5A2 in Human, Mouse, Rat samples. It is validated for Immunohistochemistry.
Specifications
| LRH-1/NR5A2 | |
| Monoclonal | |
| FITC | |
| PBS with 0.05% Sodium Azide | |
| NR5A2 | |
| Recombinant protein fragment corresponding to amino acids 1-303 of human LRH-1/NR5A2 (NP_995582) produced in E.coli. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| IgG1 |
| Immunohistochemistry | |
| OTI3H8 | |
| Immunohistochemistry | |
| Alpha-1-fetoprotein transcription factor, B1-binding factor, b1-binding factor, hepatocyte transcription factor which activates enhancer IIof hepatitis B virus, B1F, B1F2, fetoprotein-alpha 1 (AFP) transcription factor, FTFLRH1, FTZ-F1, FTZ-F1beta, HB1F, hB1F-2, hB1FCYP7A promoter-binding factor, Hepatocytic transcription factor, Liver receptor homolog 1, liver receptor homolog-1, LRH-1CPF, nuclear receptor NR5A2, nuclear receptor subfamily 5 group A member 2, nuclear receptor subfamily 5, group A, member 2 | |
| Mouse | |
| Affinity Purified | |
| Cancer, Chromatin Research, GPCR, Neuroscience, Stem Cell Markers | |
| 2494.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction