missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ LRH-1/NR5A2 Antibody (PCRP-NR5A2-1B8), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals™ NBP3-26786-100ug
This item is not returnable.
View return policy
Description
LRH-1/NR5A2 Monoclonal antibody specifically detects LRH-1/NR5A2 in Human samples. It is validated for Flow Cytometry,Immunohistochemistry (Paraffin)
Specifications
| LRH-1/NR5A2 | |
| Monoclonal | |
| 0.2 mg/mL | |
| Flow Cytometry 1-2 μg/million cells, Immunohistochemistry-Paraffin 1-2 μg/mL for 30 minutes at RT | |
| Alpha-1-fetoprotein transcription factor, B1-binding factor, b1-binding factor, hepatocyte transcription factor which activates enhancer IIof hepatitis B virus, B1F, B1F2, fetoprotein-alpha 1 (AFP) transcription factor, FTFLRH1, FTZ-F1, FTZ-F1beta, HB1F, hB1F-2, hB1FCYP7A promoter-binding factor, Hepatocytic transcription factor, Liver receptor homolog 1, liver receptor homolog-1, LRH-1CPF, nuclear receptor NR5A2, nuclear receptor subfamily 5 group A member 2, nuclear receptor subfamily 5, group A, member 2 | |
| Recombinant human LRH-1/NR5A2 protein | |
| 100 μg | |
| Cancer, Chromatin Research, Neuroscience, Stem Cell Markers | |
| 2494 | |
| Store at 4°C. | |
| IgG2b |
| Flow Cytometry, Immunohistochemistry (Paraffin) | |
| PCRP-NR5A2-1B8 | |
| Unconjugated | |
| 10 mM PBS with 0.05% BSA | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction