missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LRIG1 Antibody (789211), PE/Atto594, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB7498PEATT594
This item is not returnable.
View return policy
Description
LRIG1 Monoclonal antibody specifically detects LRIG1 in Human samples. It is validated for Flow Cytometry
Specifications
| LRIG1 | |
| Monoclonal | |
| PE/ATTO 594 | |
| PBS | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry | |
| 789211 | |
| Flow Cytometry | |
| D6Bwg0781e, Img, LIG-1 | |
| HEK293 human embryonic kidney cell line transfected with human LRIG1, Ala35-Ser779, Accession # Q96JA1 | |
| 0.1 mL | |
| Cancer | |
| 26018 | |
| Store at 4C in the dark. Do not freeze. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction