missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LRP-1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-62753
This item is not returnable.
View return policy
Description
LRP-1 Polyclonal specifically detects LRP-1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| LRP-1 | |
| Polyclonal | |
| Unconjugated | |
| PBS pH 7.4, 50% glycerol with 0.09% Sodium Azide | |
| LRP1 | |
| Synthetic peptide from the N-terminal of Human Low-density lipoprotein receptor-related protein 1 (aa. 160-260) | |
| 100 μg | |
| 4035 | |
| Human, Mouse, Rat | |
| IgG |
| Western Blot, Immunohistochemistry | |
| 1mg/mL | |
| Western Blot 1:1000, Immunohistochemistry 1:50 | |
| A2MR, Alpha-2-Macroglobulin Receptor, APOER, Apolipoprotein E receptor, APR, CD91, CD91 antigen, EC 1.1.1.94, EC 3.4.21.9, IGFBP3R, low density lipoprotein receptor-related protein 1, LRP, LRP-1, LRP1A, prolow-density lipoprotein receptor-related protein 1, TbetaR-V/LRP-1/IGFBP-3 receptor, TGFBR5, type V tgf-beta receptor | |
| Rabbit | |
| Affinity Purified | |
| Primary | |
| Detects 80 kDa. Band at 80 kDa is a membrane spanning subunit. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction