missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LRP-4 Antibody (S207-27), DyLight 488, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-22416G
This item is not returnable.
View return policy
Description
LRP-4 Monoclonal specifically detects LRP-4 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.
Specifications
| LRP-4 | |
| Monoclonal | |
| DyLight 488 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| LRP4 | |
| Fusion protein amino acids 26- 350 (extracellular N-terminus) of mouse LTP4 | |
| 0.1 mL | |
| Primary | |
| Detects the extracellular domain of LRP4 approx 215kDa. Degradation product can be seen at approx 150kDa. | |
| Store at 4C in the dark. | |
| IgG2a |
| Western Blot, Immunocytochemistry, Immunofluorescence | |
| S207-27 | |
| Western Blot, Immunocytochemistry/Immunofluorescence | |
| CLSS, KIAA0816, low density lipoprotein receptor-related protein 4, low-density lipoprotein receptor-related protein 4, LRP-4, MEGF7LRP10, Multiple epidermal growth factor-like domains 7 | |
| Mouse | |
| Protein G purified | |
| Stem Cell Markers | |
| 4038 | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction