Learn More
Invitrogen™ LRP12 Polyclonal Antibody
GREENER_CHOICE
Rabbit Polyclonal Antibody
Brand:  Invitrogen™ PA550233
Product Code. 15644165
  • € 530.00 / 100µL
Description
The antibody detects endogenous levels of total LRP12 protein.
Members of the LDL receptor gene family, including LDLR (low density lipoprotein receptor), LRP1 (low density lipoprotein related protein), Megalin (also designated GP330), VLDLR (very low density lipoprotein receptor) and ApoER2 are characterized by a cluster of cysteine-rich class A repeats, epidermal growth factor (EGF)-like repeats, YWTD repeats and an O-linked sugar domain. LRP12, also designated ST7, is a member of the LDLR family that is thought to be involved in the internalization of lipophilic molecules and/or signal transduction. LRP12 has been shown to interact with RACK1, MADHIP and Integrin beta-1-binding protein 3. It is widely expressed in heart, skeletal muscle, brain, lung, placenta and pancreas. Overexpression of LRP12 may be associated with oral tumors, therefore implicating LRP12 as a potential oncogene.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
LRP12
Polyclonal
Unconjugated
LRP12
AI848829; C820005L12Rik; LDL receptor related protein 12; LDLR-related protein 12; low density lipoprotein receptor-related protein 12; low density lipoprotein-related protein 12; low-density lipoprotein receptor-related protein 12; LRP12; LRP-12; ST7; Suppressor of tumorigenicity 7 protein
Rabbit
Antigen affinity chromatography
RUO
29967
-20°C
Liquid
Immunohistochemistry (Paraffin), Western Blot
0.8 mg/mL
PBS with 40% glycerol and 0.05% sodium azide; pH 7.4
Q9Y561
LRP12
Fusion protein corresponding to a region derived from internal residues of human low density lipoprotein receptor-related protein 12.