missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ LRP15 Monoclonal Antibody (758315)
Mouse Monoclonal Antibody
Brand: Invitrogen™ MA561848
This item is not returnable.
View return policy
Description
Reconstitute with sterile PBS to a final concentration of 0.5 mg/mL.
The protein encoded by this gene is a tumor suppressor, with lowered expression levels found in gastric, renal, colorectal, lung, and breast cancer tissues. The promoter of this gene is frequently hypermethylated in these cancer tissues, although the hypermethylation does not appear to be the cause of the reduced expression of this gene. Several transcript variants encoding the same protein have been found for this gene.
Specifications
| LRP15 | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with trehalose and no preservative | |
| Q96PB8 | |
| LRRC3B | |
| Chinese hamster ovary cell line CHO-derived recombinant human LRRC3B, Phe28-Ala205. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Western Blot | |
| 758315 | |
| Unconjugated | |
| LRRC3B | |
| BC019794; leucine rich repeat containing 3B; leucine-rich repeat protein LRP15; Leucine-rich repeat-containing protein 3B; LRP15; LRRC3B; RGD1311897; UNQ195/PRO221 | |
| Mouse | |
| Protein A/G | |
| RUO | |
| 116135 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction