missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LRP2 Antibody (545818), Alexa Fluor™ 488, R&D Systems™
Mouse Monoclonal Antibody
Brand: Novus Biologicals MAB95781AF488
This item is not returnable.
View return policy
Description
LRP2 Monoclonal specifically detects LRP2 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| LRP2 | |
| Monoclonal | |
| Alexa Fluor 488 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| LRP2 | |
| Mouse myeloma cell line NS0-derived partial recombinant human Megalin/LRP2 protein (amino acids 3510-3964) | |
| 0.1 mL | |
| Lipid and Metabolism | |
| Human | |
| IgG1 |
| Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| 545818 | |
| Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin | |
| calcium sensor protein, DBSlow-density lipoprotein receptor-related protein 2, EC 1.1.2.3, EC 3.4.21.9, Glycoprotein 330, GP330, Heymann nephritis antigen homolog, low density lipoprotein receptor-related protein 2, LRP-2, megalin | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction