missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LRP2 Antibody [Janelia Fluor™ 635], Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP3-21426JF635
This item is not returnable.
View return policy
Description
LRP2 Polyclonal antibody specifically detects LRP2 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| LRP2 | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry (Paraffin) | |
| Janelia Fluor 635 | |
| calcium sensor protein, DBSlow-density lipoprotein receptor-related protein 2, EC 1.1.2.3, EC 3.4.21.9, Glycoprotein 330, GP330, Heymann nephritis antigen homolog, low density lipoprotein receptor-related protein 2, LRP-2, megalin | |
| Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human LRP2 | |
| 0.1 mL | |
| Lipid and Metabolism | |
| 4036 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction