missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LRP2 Antibody (SR1448), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-21611-50ul
This item is not returnable.
View return policy
Description
LRP2 Monoclonal antibody specifically detects LRP2 in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
| LRP2 | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot | |
| SR1448 | |
| Western Blot 1:500-1:2000 | |
| calcium sensor protein, DBSlow-density lipoprotein receptor-related protein 2, EC 1.1.2.3, EC 3.4.21.9, Glycoprotein 330, GP330, Heymann nephritis antigen homolog, low density lipoprotein receptor-related protein 2, LRP-2, megalin | |
| A synthesized peptide derived from human LRP2 (Uniprot #: P98164) | |
| 50 μg | |
| Lipid and Metabolism | |
| 4036 | |
| Store at -20° C. Avoid freeze/thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction