Learn More
Invitrogen™ LRP4 Monoclonal Antibody (N207/27), FITC
GREENER_CHOICE
Mouse Monoclonal Antibody
Brand:  Invitrogen™ MA545577
Product Code. 17827312
  • € 467.00 / 100µg
Description
1 μg/mL of MA5-45577 was sufficient for detection of LRP4 in 20 μg of rat brain lysate by colorimetric immunoblot analysis using Goat anti-mouse IgG:HRP as the secondary antibody. Detects the extracellular domain of LRP4 approximately 215kDa. Degradation product can be seen at approximately 150kDa. This antibody was formerly sold as clone S207-27.
This gene encodes a member of the low-density lipoprotein receptor-related protein family. The encoded protein may be a regulator of Wnt signaling. Mutations in this gene are associated with Cenani-Lenz syndrome.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
LRP4
Monoclonal
1 mg/mL
PBS, 9.09mM sodium bicarbonate with 640.91mM DMSO, 136.36mM ethanolamine and no preservative; pH 7.4
O75096, Q8VI56, Q9QYP1
LRP4
Fusion protein amino acids 26- 350 (extracellular N-terminus) of mouse LTP4.
100 μg
Primary
Human, Mouse, Rat
Antibody
IgG2a
Western Blot, Immunocytochemistry
N207/27
FITC
LRP4
6430526J12Rik; CLSS; CMS17; Corin; D230026E03; Kiaa0816; LDL receptor related protein 4; LDLR; LDLR dan; low density lipoprotein receptor-related protein 4; low-density lipoprotein receptor-related protein 4; LRP10; Lrp4; LRP-4; mdig; Megf7; multiple epidermal growth factor-like domains 7; SOST2
Mouse
Protein G
RUO
228357, 4038, 83469
4°C, store in dark