missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LRP6, Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit Polyclonal Antibody
Brand: Abnova PAB12632.100ug
This item is not returnable.
View return policy
Description
Sequence: KGTYFSpecifications
| LRP6 | |
| Polyclonal | |
| Western Blot (0.1-1 ug/mL) ELISA (0.01-0.1 ug/mL) Immunoprecipitation (2-5 ug/mL) The optimal working dilution should be determined by the end user. | |
| low density lipoprotein receptor-related protein 6 | |
| LRP6 | |
| Synthetic phosphopeptide corresponding to residues surrounding T1479 of human LRP6. | |
| RUO | |
| 4040 | |
| Human, Mouse, Rat | |
| Antibody |
| Dot Blot, ELISA, Immunoprecipitation, Western Blot | |
| Unconjugated | |
| In TBS, pH 7.2 (BSA, 10% Proclin300) | |
| ADCAD2/FLJ90062/FLJ90421 | |
| Rabbit | |
| 100 μg | |
| Primary | |
| This antibody recognizes ∼180 KDa of human LRP6. | |
| Store at 4°C. For long term storage store at -20°C to -80°C. Aliquot to avoid repeated freezing and thawing. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction