missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LRRC16B Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-88687
This item is not returnable.
View return policy
Beskrivning
LRRC16B Polyclonal specifically detects LRRC16B in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifikationer
| LRRC16B | |
| Polyclonal | |
| Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
| C14orf121, CARMIL3, crml-1, leucine rich repeat containing 16B, leucine-rich repeat-containing protein 16B | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 90668 | |
| Human | |
| IgG |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| LRRC16B | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:CDYNGLHCREEVQWDVDTIYHAEDNREFNLLDFSHLESRDLALMVAALAYNQWFTKLYCKDLRLGSEVLEQVLHTLSKSGSLEELVLDNAGLKTDFVQKLAGVFGENGSCVLHALTL | |
| 0.1 mL | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
For Research Use Only
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering