missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LRRC17 Antibody, Alexa Fluor™ 350, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-99220AF350
This item is not returnable.
View return policy
Description
LRRC17 Polyclonal antibody specifically detects LRRC17 in Human samples. It is validated for Western Blot
Specifications
| LRRC17 | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| 10234 | |
| Store at 4°C in the dark. | |
| IgG |
| Western Blot | |
| Alexa Fluor 350 | |
| 37 kDa leucine-rich repeat (LRR) protein, leucine rich repeat containing 17, leucine-rich repeat-containing protein 17, p37NB, P37NBH_RG318M05.3 | |
| Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of the Human LRRC17. | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction