missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LRRC25 Mouse anti-Human, Clone: OTI1D9, liquid, TrueMAB™
Mouse Monoclonal Antibody
Brand: Origene Technologies TA504883
This item is not returnable.
View return policy
Description
May be involved in the activation of cells of innate and acquired immunity.Specifications
| LRRC25 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q8N386 | |
| LRRC25 | |
| Full length human recombit protein of human LRRC25 produced in HEK293T cell. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
| OTI1D9 | |
| Unconjugated | |
| LRRC25 | |
| MAPA | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 126364 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction