missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ LRRC29 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5116268
This item is not returnable.
View return policy
Description
LRRC29 Polyclonal Antibody for Western Blot
Specifications
| LRRC29 | |
| Polyclonal | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| Q8WV35 | |
| LRRC29 | |
| KLH conjugated synthetic peptide between 32-60 amino acids from the N-terminal region of human LRRC29. | |
| 400 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Western Blot | |
| Unconjugated | |
| LRRC29 | |
| A630024J02Rik; FBL9; F-box and leucine-rich repeat protein 9; F-box protein FBL9; F-box/LRR-repeat protein 9; FBXL9; FBXL9P; leucine rich repeat containing 29; leucine-rich repeat-containing protein 29; LRRC29 | |
| Rabbit | |
| Antigen Affinity Chromatography, Protein A | |
| RUO | |
| 26231 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction