missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LRRC32/GARP Antibody (725226) [FITC], Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals FAB62291F
This item is not returnable.
View return policy
Description
LRRC32/GARP Monoclonal antibody specifically detects LRRC32/GARP in Mouse samples. It is validated for Flow Cytometry, CyTOF
Specifications
| LRRC32/GARP | |
| Monoclonal | |
| FITC | |
| PBS | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 2615 | |
| Store at 4°C in the dark. | |
| IgG1 |
| Flow Cytometry, CyTOF | |
| 725226 | |
| Flow Cytometry, CyTOF-ready | |
| D11S833Eleucine-rich repeat-containing protein 32, GARPGarpin, Glycoprotein A repetitions predominantgarpin, leucine rich repeat containing 32 | |
| Chinese hamster ovary cell line CHO-derived recombinant mouse LRRC32/GARP, Ile18-Asn628, Accession # NP_001106850 | |
| 0.1 mL | |
| Primary | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction