missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LRRC35 Antibody (1B10), CoraFluor™ 1, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-59417CL1
This item is not returnable.
View return policy
Description
LRRC35 Monoclonal antibody specifically detects LRRC35 in Human samples. It is validated for Flow Cytometry,ELISA,Western Blot,Immunocytochemistry/Immunofluorescence
Specifications
| LRRC35 | |
| Monoclonal | |
| CoraFluor 1 | |
| tubulin-specific chaperone cofactor E-like protein, El, LRRC35, tubulin folding cofactor E-like | |
| Recombinant human LRRC35 (1-424aa) purified from E. coli | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry, ELISA, Western Blot, Immunocytochemistry/Immunofluorescence | |
| 1B10 | |
| PBS | |
| Mouse | |
| Protein A purified | |
| RUO | |
| 219899 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG2b κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction