missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LRRC35 Mouse anti-Human, Mouse, Clone: 1B10, Novus Biologicals™
Mouse Monoclonal Antibody
€ 294.00 - € 486.00
Specifications
| Antigen | LRRC35 |
|---|---|
| Clone | 1B10 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:1000, Flow Cytometry 1:10-1:1027, ELISA, Immunocytochemistry/Immunofluorescence 1:100 |
| Applications | Western Blot, Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18212965
|
Novus Biologicals
NBP2-59417-50UL |
50 μL |
€ 294.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18262001
|
Novus Biologicals
NBP2-59417-100UL |
100 μL |
€ 486.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
LRRC35 Monoclonal specifically detects LRRC35 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence.Specifications
| LRRC35 | |
| 1 mg/mL | |
| Western Blot, Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| Mouse | |
| Human, Mouse | |
| tubulin-specific chaperone cofactor E-like protein, El, LRRC35, tubulin folding cofactor E-like | |
| TBCEL | |
| IgG2b κ | |
| Protein A purified |
| 1B10 | |
| Western Blot 1:1000, Flow Cytometry 1:10-1:1027, ELISA, Immunocytochemistry/Immunofluorescence 1:100 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Liquid. In PBS (pH 7.4), 10% Glycerol with 0.02% Sodium Azide | |
| 219899 | |
| Recombinant human TBCEL (1-424aa) purified from E. coli | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title