Learn More
LRRC37A Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand:  Novus Biologicals NBP2-46690-25ul
 View more versions of this product
Product Code. 18690546
  • € 415.00 / 25µL
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.
Description
Description
LRRC37A Polyclonal antibody specifically detects LRRC37A in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Spécification
Spécification
LRRC37A
Polyclonal
Immunohistochemistry 1:500 - 1:1000, Immunohistochemistry-Paraffin 1:500 - 1:1000
A6NMS7
Rabbit
Immunogen affinity purified
RUO
9884
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
IgG
Immunohistochemistry, Immunohistochemistry (Paraffin)
Unconjugated
PBS (pH 7.2), 40% Glycerol
Leucine Rich Repeat Containing 37A, Leucine-Rich Repeat-Containing Protein 37A, LRRC37, LRRC37A1
This antibody was developed against Recombinant Protein corresponding to amino acids: PGQPPEHHEVTVSPPGHHQTHHLASPSVSVKPPDVQLTIAAEPSAEVGTSLVHQEATTRLSGSGNDVEPPAIQHGGPPLLPESSEE