missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LRRC39 Antibody (OTI2D9), Alexa Fluor™ 405, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-72533AF405
This item is not returnable.
View return policy
Description
LRRC39 Monoclonal specifically detects LRRC39 in Human, Mouse, Rat samples. It is validated for Western Blot.
Specifications
| LRRC39 | |
| Monoclonal | |
| Alexa Fluor 405 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| LRRC39 | |
| Full length recombinant protein of human LRRC39 (NP_653221) produced in HEK293T cells. | |
| 0.1 mL | |
| 127495.0 | |
| Store at 4°C in the dark. |
| Western Blot | |
| OTI2D9 | |
| Western Blot | |
| Densin hlg, DKFZp313O1122, leucine rich repeat containing 39, leucine-rich repeat-containing protein 39, MGC14816 | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human, Mouse, Rat | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction