missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LRRC3B/LRP15 Antibody, DyLight 650, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-26069C
This item is not returnable.
View return policy
Description
LRRC3B/LRP15 Polyclonal antibody specifically detects LRRC3B/LRP15 in Human samples. It is validated for ELISA
Specifications
| LRRC3B/LRP15 | |
| Polyclonal | |
| ELISA | |
| leucine rich repeat containing 3B, leucine-rich repeat-containing protein 3B, LRP15Leucine-rich repeat protein LRP15, MGC102927 | |
| Rabbits immunized with purified, recombinant Human LRRC3B/LRP15 (Accession#: NP_443185.1, Met1-Tyr204) | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| ELISA | |
| DyLight 650 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| 116135 | |
| Store at 4C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction