Learn More
LRRC41 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 20457-1-AP-150UL
 View more versions of this product
Product Code. 16820005
  • € 410.00 / 150µL
Description
Chromosome 1 is the largest human chromosome spanning about 260 million base pairs and making up 8% of the human genome. There are about 3, 000 genes on chromosome 1, and considering the great number of genes there are also a large number of diseases associated with chromosome 1. Notably, the rare aging disease Hutchinson-Gilford progeria is associated with the LMNA gene which encodes lamin A. When defective, the LMNA gene product can build up in the nucleus and cause characteristic nuclear blebs. The mechanism of rapidly enhanced aging is unclear and is a topic of continuing exploration. The MUTYH gene is located on chromosome 1 and is partially responsible for familial adenomatous polyposis.
Specifications
Specifications
LRRC41
Polyclonal
Unconjugated
LRRC41
LRRC41, MUF1, PP7759, Protein Muf1, RP4 636H5.2
Rabbit
Antigen Affinity Chromatography
RUO
10489, 230654, 362566
-20°C
Liquid
Immunohistochemistry (Paraffin), Western Blot
0.13 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q15345, Q5M9H1, Q8K1C9
Lrrc41
LRRC41 Fusion Protein Ag14266
150 μL
Primary
Human, Mouse, Rat