missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ LRRC43 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA561553
This item is not returnable.
View return policy
Description
Immunogen sequence: LPAVDSPLSA KKGKGEKDKK GKEKDRTGKG EKEPAKEWKV LKKKKEPPKE LRQDPPILQV LGRGLVILEP LLAGEPLVST VCNFGVVRTL TSDRLTLARD SKKIKKVAK Highest antigen sequence identity to the following orthologs: Mouse - 66%, Rat - 65%.
LRRC43 is a protein coding gene.
Specifications
| LRRC43 | |
| Polyclonal | |
| Unconjugated | |
| LRRC43 | |
| leucine rich repeat containing 43; leucine-rich repeat-containing protein 43; LRRC43 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 254050 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.4 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q8N309 | |
| LRRC43 | |
| Recombinant protein corresponding to Human LRRC43. Recombinant protein control fragment (Product #RP-97843). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction