missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ LRRC48 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA546526
This item is not returnable.
View return policy
Description
Peptide sequence: NIFEYGLKQQ EKRKTELDTF SECVREAIQE NQEQGKRKIA KFEEKHLSLF Sequence homology: Cow: 93%; Dog: 86%; Guinea Pig: 85%; Horse: 93%; Human: 100%; Rabbit: 92%; Rat: 86%.
Component of the nexin-dynein regulatory complex (N-DRC) a key regulator of ciliary/flagellar motility which maintains the alignment and integrity of the distal axoneme and regulates microtubule sliding in motile axonemes. [UniProt].
Specifications
| LRRC48 | |
| Polyclonal | |
| Unconjugated | |
| DRC3 | |
| CFAP134; DRC3; Dynein regulatory complex subunit 3; dynein regulatory complex subunit 3; LOW QUALITY PROTEIN: dynein regulatory complex subunit 3; leucine rich repeat containing 48; leucine-rich repeat-containing protein 48; Lrrc48; RGD1309150 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 83450 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q9H069 | |
| DRC3 | |
| Synthetic peptide directed towards the C-terminal of human LRRC48. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction