missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ LRRC49 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA570270
This item is not returnable.
View return policy
Description
This target displays homology in the following species: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%.
LRRC49 is a protein coding gene.
Specifications
| LRRC49 | |
| Polyclonal | |
| Unconjugated | |
| LRRC49 | |
| AI430817; AW050057; D430025H09Rik; leucine rich repeat containing 49; leucine-rich repeat-containing protein 49; Lrrc49; p79; PGs4; RGD1309466; Tubulin polyglutamylase complex subunit 4 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 54839 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q8IUZ0 | |
| LRRC49 | |
| synthetic peptide directed towards the N terminal region of human LRRC49. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction