missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LRRC50 Antibody (OTI2C4), CoraFluor™ 1, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP2-72536CL1
This item is not returnable.
View return policy
Description
LRRC50 Monoclonal antibody specifically detects LRRC50 in Canine,Human,Monkey,Mouse,Primate,Rat samples. It is validated for Flow Cytometry,Western Blot,CyTOF
Specifications
| LRRC50 | |
| Monoclonal | |
| CoraFluor 1 | |
| CILD13, DKFZp434A119, FLJ25330, leucine rich repeat containing 50, ODA7leucine-rich repeat-containing protein 50, outer row dynein assembly 7 homolog | |
| Full length human recombinant protein of human LRRC50(NP_848547) produced in HEK293T cell. | |
| 0.1 mL | |
| Cell Biology, Cytoskeleton Markers, Signal Transduction | |
| 123872 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG1 |
| Flow Cytometry, Western Blot, CyTOF | |
| OTI2C4 | |
| PBS | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Canine, Human, Monkey, Mouse, Primate | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction