missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LRRC50 Antibody (OTI2C4), HRP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-72536H
This item is not returnable.
View return policy
Description
LRRC50 Monoclonal specifically detects LRRC50 in Human, Mouse, Rat, Canine, Primate, Monkey samples. It is validated for Western Blot.
Specifications
| LRRC50 | |
| Monoclonal | |
| HRP | |
| PBS with No Preservative | |
| DNAAF1 | |
| Full length recombinant protein of human LRRC50 (NP_848547) produced in HEK293T cells. | |
| 0.1 mL | |
| 123872.0 | |
| Store at 4°C in the dark. |
| Western Blot | |
| OTI2C4 | |
| Western Blot | |
| CILD13, DKFZp434A119, FLJ25330, leucine rich repeat containing 50, ODA7leucine-rich repeat-containing protein 50, outer row dynein assembly 7 homolog | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human, Mouse, Rat, Canine, Primate, Monkey | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction