Learn More
LRRC50 Monoclonal Antibody (OTI2C4), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA504546
This item is not returnable.
View return policy
Description
The protein encoded by this gene is cilium-specific and is required for the stability of the ciliary architecture. It is involved in the regulation of microtubule-based cilia and actin-based brush border microvilli. Mutations in this gene are associated with primary ciliary dyskinesia-13.Specifications
| LRRC50 | |
| Monoclonal | |
| 2.74 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q6AYH9, Q8NEP3, Q9D2H9 | |
| Dnaaf1 | |
| Full length human recombit protein of human LRRC50 produced in HEK293T cell. | |
| 100 μL | |
| Primary | |
| Canine, Human, Mouse, Rat | |
| Antibody | |
| IgG1 |
| Flow Cytometry, Western Blot | |
| OTI2C4 | |
| Unconjugated | |
| DNAAF1 | |
| CILD13, LRRC50, ODA7, swt | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 123872, 361419, 479628, 68270 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |