Learn More
LRRC50 Monoclonal Antibody (OTI2C4), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA504546S
This item is not returnable.
View return policy
Description
LRRC50 Monoclonal antibody specifically detects LRRC50 in Human samples. It is validated for Flow Cytometry, Western BlotSpecifications
| LRRC50 | |
| Monoclonal | |
| 2.74 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q8TBE9 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 140838 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Flow Cytometry, Western Blot | |
| OTI2C4 | |
| Unconjugated | |
| NANP | |
| 4930457P18Rik; CILD13; Dnaaf1; dynein (axonemal) assembly factor 1; dynein assembly factor 1, axonemal; dynein axonemal assembly factor 1; dynein, axonemal assembly factor 1; dynein, axonemal, assembly factor 1; hypothetical protein LOC386722; hypothetical protein LOC523187; leucine rich repeat containing 50; leucine-rich repeat-containing protein 50; LLRC50; LOW QUALITY PROTEIN: dynein assembly factor 1, axonemal; lrrc50; ODA7; outer row dynein assembly 7 homolog; RGD1310542; switch hitter; swt; testicular tissue protein Li 110; unm tm317b; zgc:56169 | |
| Full length human recombinant protein of human NANP produced in HEK293T cell. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |