Learn More
CD68 Monoclonal Antibody (OTI4D8), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand:  Origene Technologies TA802983S
Encompass_Preferred
 View more versions of this product
Product Code. 17581853
  • POA / 30µL
This item requires a price update prior to being ordered.
Request a Price for this item here.
View alternative products »
Description
Description
CD68 Monoclonal antibody specifically detects CD68 in Human samples. It is validated for Western Blot
Specifications
Specifications
CD68
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
P34810
Mouse
Affinity chromatography
RUO
968
-20° C, Avoid Freeze/Thaw Cycles
IgG2a
Western Blot
OTI4D8
Unconjugated
Cd68
CD68; CD68 antigen; CD68 molecule; GP110; Lamp4; macrophage antigen CD68; macrosialin; Scard1; scavenger receptor class D, member 1; similar to CD68 antigen
Human recombinant protein fragment corresponding to amino acids 22-319 of human CD68 produced in SF9 cell.
30 μL