missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ LRRC50 Polyclonal Antibody
GREENER_CHOICE

Product Code. 17161864 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17161864 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17161864 Supplier Invitrogen™ Supplier No. PA5114006

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: TELRCLFLQM NLLRKIENLE PLQKLDALNL SNNYIKTIEN LSCLPVLNTL For short term use, store at 2-8°C up to 1 week. For long term storage, store at -20°C in small aliquots to prevent freeze-thaw cycles. Predicted homology: Cow: 100%; Dog: 93%; Guinea Pig: 100%; Horse: 93%; Human: 100%; Mouse: 100%; Rat: 100%.

The protein encoded by this gene is cilium-specific and is required for the stability of the ciliary architecture. It is involved in the regulation of microtubule-based cilia and actin-based brush border microvilli. Mutations in this gene are associated with primary ciliary dyskinesia-13.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LRRC50
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene DNAAF1
Gene Accession No. Q8NEP3
Gene Alias 4930457P18Rik; CILD13; Dnaaf1; dynein (axonemal) assembly factor 1; dynein assembly factor 1, axonemal; dynein axonemal assembly factor 1; dynein, axonemal assembly factor 1; dynein, axonemal, assembly factor 1; hypothetical protein LOC386722; hypothetical protein LOC523187; leucine rich repeat containing 50; leucine-rich repeat-containing protein 50; LLRC50; LOW QUALITY PROTEIN: dynein assembly factor 1, axonemal; lrrc50; ODA7; outer row dynein assembly 7 homolog; RGD1310542; switch hitter; swt; testicular tissue protein Li 110; unm tm317b; zgc:56169
Gene Symbols DNAAF1
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N terminal region of human LRRC50 (aa 151-200).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 123872
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.