missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ LRRC50 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA532025
This item is not returnable.
View return policy
Description
Recommended positive controls: mouse testis. Predicted reactivity: Mouse (87%), Rat (86%), Bovine (88%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
The protein encoded by this gene is cilium-specific and is required for the stability of the ciliary architecture. It is involved in the regulation of microtubule-based cilia and actin-based brush border microvilli. Mutations in this gene are associated with primary ciliary dyskinesia-13.
Specifications
| LRRC50 | |
| Polyclonal | |
| Unconjugated | |
| DNAAF1 | |
| 4930457P18Rik; CILD13; Dnaaf1; dynein (axonemal) assembly factor 1; dynein assembly factor 1, axonemal; dynein axonemal assembly factor 1; dynein, axonemal assembly factor 1; dynein, axonemal, assembly factor 1; hypothetical protein LOC386722; hypothetical protein LOC523187; leucine rich repeat containing 50; leucine-rich repeat-containing protein 50; LLRC50; LOW QUALITY PROTEIN: dynein assembly factor 1, axonemal; lrrc50; ODA7; outer row dynein assembly 7 homolog; RGD1310542; switch hitter; swt; testicular tissue protein Li 110; unm tm317b; zgc:56169 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 123872, 68270 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 0.96 mg/mL | |
| 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 | |
| Q8NEP3, Q9D2H9 | |
| DNAAF1 | |
| Recombinant protein encompassing a sequence within the N-terminus region of human LRRC50. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction